Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611408 MIM: 615678 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCA5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LCA5 - LCA5, lebercilin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3BGRL2 - SH3 domain binding glutamate rich protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031469.3 | Intron | NP_113657.1 | ||||
XM_011536165.2 | Intron | XP_011534467.1 | ||||
XM_011536282.2 | Intron | XP_011534584.1 | ||||
XM_017011344.1 | Intron | XP_016866833.1 | ||||
XM_017011345.1 | Intron | XP_016866834.1 |