Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131235 MIM: 608373 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDELR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KDELR1 - KDEL endoplasmic reticulum protein retention receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNGR4 - synaptogyrin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012451.3 | 253 | Intron | NP_036583.2 | |||
XM_005258693.2 | 253 | Intron | XP_005258750.1 | |||
XM_005258694.2 | 253 | Intron | XP_005258751.1 | |||
XM_011526686.2 | 253 | Intron | XP_011524988.1 | |||
XM_017026541.1 | 253 | UTR 5 | XP_016882030.1 |
TMEM143 - transmembrane protein 143 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |