Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603617 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC24A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SLC24A1 - solute carrier family 24 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254740.1 | 370 | Intron | NP_001241669.1 | |||
NM_001301031.1 | 370 | Missense Mutation | ACT,TCT | T,S 37 | NP_001287960.1 | |
NM_001301032.1 | 370 | Missense Mutation | ACT,TCT | T,S 37 | NP_001287961.1 | |
NM_001301033.1 | 370 | Missense Mutation | ACT,TCT | T,S 37 | NP_001287962.1 | |
NM_004727.2 | 370 | Missense Mutation | ACT,TCT | T,S 37 | NP_004718.1 | |
XM_005254778.3 | 370 | Missense Mutation | ACT,TCT | T,S 37 | XP_005254835.1 | |
XM_005254781.3 | 370 | Missense Mutation | ACT,TCT | T,S 37 | XP_005254838.1 | |
XM_006720768.3 | 370 | Missense Mutation | ACT,TCT | T,S 37 | XP_006720831.1 | |
XM_011522219.1 | 370 | Missense Mutation | ACT,TCT | T,S 37 | XP_011520521.1 | |
XM_011522220.2 | 370 | Missense Mutation | ACT,TCT | T,S 37 | XP_011520522.1 | |
XM_011522221.2 | 370 | Missense Mutation | ACT,TCT | T,S 37 | XP_011520523.1 | |
XM_011522222.2 | 370 | Missense Mutation | ACT,TCT | T,S 37 | XP_011520524.1 | |
XM_011522226.2 | 370 | Missense Mutation | ACT,TCT | T,S 37 | XP_011520528.1 | |
XM_017022724.1 | 370 | Missense Mutation | ACT,TCT | T,S 37 | XP_016878213.1 | |
XM_017022725.1 | 370 | Missense Mutation | ACT,TCT | T,S 37 | XP_016878214.1 |
Set Membership: |
HapMap Validated |