Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCAGATTAGAAAATGGCAGGG[C/T]GCTGGCACCCCGCTTCATCACACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600489 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFATC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFATC1 - nuclear factor of activated T-cells 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278669.1 | Intron | NP_001265598.1 | ||||
NM_001278670.1 | Intron | NP_001265599.1 | ||||
NM_001278672.1 | Intron | NP_001265601.1 | ||||
NM_001278673.1 | Intron | NP_001265602.1 | ||||
NM_001278675.1 | Intron | NP_001265604.1 | ||||
NM_006162.4 | Intron | NP_006153.2 | ||||
NM_172387.2 | Intron | NP_765975.1 | ||||
NM_172388.2 | Intron | NP_765976.1 | ||||
NM_172389.2 | Intron | NP_765977.1 | ||||
NM_172390.2 | Intron | NP_765978.1 | ||||
XM_017025783.1 | Intron | XP_016881272.1 |