Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAAATAAATAAAAGCAGAGGGAT[A/T]TCTGAGACGCAGAGACACAGGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615585 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NECAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NECAB2 - N-terminal EF-hand calcium binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC38A8 - solute carrier family 38 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080442.2 | Intron | NP_001073911.1 | ||||
XM_017022946.1 | Intron | XP_016878435.1 |
Set Membership: |
HapMap |