Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603684 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPG PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LIPG - lipase G, endothelial type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308006.1 | 50 | Intron | NP_001294935.1 | |||
NM_006033.3 | 50 | Intron | NP_006024.1 | |||
XM_005258390.1 | 50 | UTR 5 | XP_005258447.1 | |||
XM_011526265.2 | 50 | Intron | XP_011524567.1 | |||
XM_011526267.1 | 50 | UTR 5 | XP_011524569.1 | |||
XM_017026095.1 | 50 | UTR 5 | XP_016881584.1 |
Set Membership: |
HapMap Validated |