Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACTTAACAAAGCATAAACACTC[C/T]CTCAAGGATGATCTAAACACTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613173 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEGR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NEGR1 - neuronal growth regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173808.2 | Intron | NP_776169.2 | ||||
XM_011541200.2 | Intron | XP_011539502.1 | ||||
XM_011541201.2 | Intron | XP_011539503.1 | ||||
XM_017000961.1 | Intron | XP_016856450.1 |
Set Membership: |
HapMap |