Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGGAAAGCAACAGCTGTAAGAT[T/C]TTTGTGGATCTCAAAGGTTAGGCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604752 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF267 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF267 - zinc finger protein 267 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001265588.1 | Intron | NP_001252517.1 | ||||
NM_003414.5 | Intron | NP_003405.3 | ||||
XM_017022865.1 | Intron | XP_016878354.1 | ||||
XM_017022866.1 | Intron | XP_016878355.1 |
Set Membership: |
HapMap |