Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGAGACTCCTTTTAGGATTCTTT[A/G]TCATTGAGGAGGCGCCGACTCATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605122 MIM: 605506 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUPV3L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SUPV3L1 - Suv3 like RNA helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301683.1 | Intron | NP_001288612.1 | ||||
NM_001323584.1 | Intron | NP_001310513.1 | ||||
NM_001323585.1 | Intron | NP_001310514.1 | ||||
NM_001323586.1 | Intron | NP_001310515.1 | ||||
NM_001323587.1 | Intron | NP_001310516.1 | ||||
NM_001323588.1 | Intron | NP_001310517.1 | ||||
NM_003171.4 | Intron | NP_003162.2 |
VPS26A - VPS26, retromer complex component A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |