Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGAGCAATTGAAGTTTAGGTGA[A/G]TAGTCTTGGGGAAGTTTATAGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
54 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606803 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACOT11 - acyl-CoA thioesterase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MROH7 - maestro heat like repeat family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039464.3 | Intron | NP_001034553.2 | ||||
NM_001291332.1 | Intron | NP_001278261.1 |
MROH7-TTC4 - MROH7-TTC4 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |