Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGGGTAAACCCACTTCCTGGTC[A/G]ATGGGCAAAACAACAGCTGGGGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606225 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOL9 - nucleolar protein 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAS1R1 - taste 1 receptor member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138697.3 | Intron | NP_619642.2 | ||||
NM_177540.2 | Intron | NP_803884.1 | ||||
XM_011542203.2 | Intron | XP_011540505.1 | ||||
XM_011542206.2 | Intron | XP_011540508.1 | ||||
XM_017002402.1 | Intron | XP_016857891.1 | ||||
XM_017002403.1 | Intron | XP_016857892.1 |
Set Membership: |
HapMap |