Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603768 MIM: 600104 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R12B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP1R12B - protein phosphatase 1 regulatory subunit 12B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYT2 - synaptotagmin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136504.1 | 3672 | UTR 3 | NP_001129976.1 | |||
NM_177402.4 | 3672 | UTR 3 | NP_796376.2 | |||
XM_011509192.2 | 3672 | UTR 3 | XP_011507494.1 | |||
XM_017000309.1 | 3672 | UTR 3 | XP_016855798.1 | |||
XM_017000310.1 | 3672 | UTR 3 | XP_016855799.1 | |||
XM_017000311.1 | 3672 | UTR 3 | XP_016855800.1 | |||
XM_017000312.1 | 3672 | UTR 3 | XP_016855801.1 | |||
XM_017000313.1 | 3672 | UTR 3 | XP_016855802.1 |
Set Membership: |
HapMap |