Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612003 MIM: 603208 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIGYF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GIGYF2 - GRB10 interacting GYF protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001103146.1 | 1119 | Intron | NP_001096616.1 | |||
NM_001103147.1 | 1119 | Intron | NP_001096617.1 | |||
NM_001103148.1 | 1119 | Intron | NP_001096618.1 | |||
NM_015575.3 | 1119 | Intron | NP_056390.2 |
KCNJ13 - potassium voltage-gated channel subfamily J member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172416.1 | 1119 | UTR 3 | NP_001165887.1 | |||
NM_001172417.1 | 1119 | UTR 3 | NP_001165888.1 | |||
NM_002242.4 | 1119 | UTR 3 | NP_002233.2 |
Set Membership: |
HapMap |