Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603217 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STX7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STX7 - syntaxin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326578.1 | 1594 | UTR 3 | NP_001313507.1 | |||
NM_001326579.1 | 1594 | UTR 3 | NP_001313508.1 | |||
NM_001326580.1 | 1594 | Intron | NP_001313509.1 | |||
NM_003569.2 | 1594 | UTR 3 | NP_003560.2 | |||
XM_011536178.1 | 1594 | Intron | XP_011534480.1 |
Set Membership: |
HapMap |