Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613621 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUBPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUBPL - nucleotide binding protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201573.1 | Intron | NP_001188502.1 | ||||
NM_001201574.1 | Intron | NP_001188503.1 | ||||
NM_025152.2 | Intron | NP_079428.2 | ||||
XM_011537181.2 | Intron | XP_011535483.1 | ||||
XM_011537182.2 | Intron | XP_011535484.1 | ||||
XM_011537183.2 | Intron | XP_011535485.1 | ||||
XM_011537184.2 | Intron | XP_011535486.1 | ||||
XM_017021664.1 | Intron | XP_016877153.1 | ||||
XM_017021665.1 | Intron | XP_016877154.1 | ||||
XM_017021666.1 | Intron | XP_016877155.1 | ||||
XM_017021667.1 | Intron | XP_016877156.1 |