Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAAGGGATTAAATCAAATCTTT[C/T]GGGGGAGAGCTCAGCTCTAGGCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191311 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DDR2 - discoidin domain receptor tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014796.1 | 17 | Intron | NP_001014796.1 | |||
NM_006182.2 | 17 | Intron | NP_006173.2 | |||
XM_006711344.3 | 17 | Intron | XP_006711407.1 | |||
XM_011509586.2 | 17 | UTR 5 | XP_011507888.1 | |||
XM_011509587.2 | 17 | UTR 5 | XP_011507889.1 | |||
XM_011509588.2 | 17 | Intron | XP_011507890.1 |
Set Membership: |
HapMap |