Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAGGGAATAAGCAATCTGGCAT[A/G]GCTTGGAGTCTAGTATTACGGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606221 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IKZF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IKZF3 - IKAROS family zinc finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257408.1 | 4065 | UTR 3 | NP_001244337.1 | |||
NM_001257409.1 | 4065 | UTR 3 | NP_001244338.1 | |||
NM_001257410.1 | 4065 | UTR 3 | NP_001244339.1 | |||
NM_001257411.1 | 4065 | UTR 3 | NP_001244340.1 | |||
NM_001257412.1 | 4065 | UTR 3 | NP_001244341.1 | |||
NM_001257413.1 | 4065 | UTR 3 | NP_001244342.1 | |||
NM_001257414.1 | 4065 | UTR 3 | NP_001244343.1 | |||
NM_001284514.1 | 4065 | UTR 3 | NP_001271443.1 | |||
NM_001284515.1 | 4065 | UTR 3 | NP_001271444.1 | |||
NM_001284516.1 | 4065 | UTR 3 | NP_001271445.1 | |||
NM_012481.4 | 4065 | UTR 3 | NP_036613.2 | |||
NM_183228.2 | 4065 | UTR 3 | NP_899051.1 | |||
NM_183229.2 | 4065 | UTR 3 | NP_899052.1 | |||
NM_183230.2 | 4065 | UTR 3 | NP_899053.1 | |||
NM_183231.2 | 4065 | UTR 3 | NP_899054.1 | |||
NM_183232.2 | 4065 | UTR 3 | NP_899055.1 |
Set Membership: |
Validated |