Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DENND3 - DENN domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014957.2 | Intron | NP_055772.2 | ||||
XM_005250838.4 | Intron | XP_005250895.2 | ||||
XM_005250839.2 | Intron | XP_005250896.2 | ||||
XM_005250840.4 | Intron | XP_005250897.2 | ||||
XM_005250841.3 | Intron | XP_005250898.2 | ||||
XM_005250842.4 | Intron | XP_005250899.1 | ||||
XM_011516933.1 | Intron | XP_011515235.1 | ||||
XM_011516934.2 | Intron | XP_011515236.1 | ||||
XM_011516937.1 | Intron | XP_011515239.1 | ||||
XM_011516938.2 | Intron | XP_011515240.1 | ||||
XM_011516939.2 | Intron | XP_011515241.1 | ||||
XM_011516940.2 | Intron | XP_011515242.1 | ||||
XM_011516941.2 | Intron | XP_011515243.1 | ||||
XM_017013241.1 | Intron | XP_016868730.1 | ||||
XM_017013242.1 | Intron | XP_016868731.1 | ||||
XM_017013243.1 | Intron | XP_016868732.1 |