Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600583 MIM: 600058 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TEC - tec protein tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003215.2 | 4391 | UTR 3 | NP_003206.2 | |||
XM_011513737.2 | 4391 | UTR 3 | XP_011512039.1 | |||
XM_011513741.1 | 4391 | Intron | XP_011512043.1 | |||
XM_017008569.1 | 4391 | UTR 3 | XP_016864058.1 |
TXK - TXK tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003328.2 | 4391 | Intron | NP_003319.2 | |||
XM_011513747.2 | 4391 | Intron | XP_011512049.1 | |||
XM_011513748.2 | 4391 | Intron | XP_011512050.1 | |||
XM_017008580.1 | 4391 | Intron | XP_016864069.1 | |||
XM_017008581.1 | 4391 | Intron | XP_016864070.1 | |||
XM_017008582.1 | 4391 | Intron | XP_016864071.1 |