Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCCCAACCAGATAGCCTCCAGC[A/G]CCACAGCCTTTTACCTGCAGCTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD2B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MFSD2B - major facilitator superfamily domain containing 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080473.1 | 1253 | Missense Mutation | ACC,GCC | T,A 60 | NP_001073942.1 | |
XM_005264301.4 | 1253 | Missense Mutation | ACC,GCC | T,A 60 | XP_005264358.1 | |
XM_017004063.1 | 1253 | Missense Mutation | ACC,GCC | T,A 60 | XP_016859552.1 | |
XM_017004064.1 | 1253 | Missense Mutation | ACC,GCC | T,A 60 | XP_016859553.1 | |
XM_017004065.1 | 1253 | Missense Mutation | ACC,GCC | T,A 60 | XP_016859554.1 | |
XM_017004066.1 | 1253 | Missense Mutation | ACC,GCC | T,A 60 | XP_016859555.1 | |
XM_017004067.1 | 1253 | Missense Mutation | ACC,GCC | T,A 60 | XP_016859556.1 | |
XM_017004068.1 | 1253 | Missense Mutation | ACC,GCC | T,A 60 | XP_016859557.1 | |
XM_017004069.1 | 1253 | Missense Mutation | ACC,GCC | T,A 60 | XP_016859558.1 | |
XM_017004070.1 | 1253 | Missense Mutation | ACC,GCC | T,A 60 | XP_016859559.1 |