Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604935 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMRT2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DMRT2 - doublesex and mab-3 related transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130865.2 | Intron | NP_001124337.1 | ||||
NM_006557.6 | Intron | NP_006548.1 | ||||
NM_181872.4 | Intron | NP_870987.2 | ||||
XM_011517687.1 | Intron | XP_011515989.1 | ||||
XM_011517690.2 | Intron | XP_011515992.1 | ||||
XM_011517694.2 | Intron | XP_011515996.1 | ||||
XM_017014213.1 | Intron | XP_016869702.1 | ||||
XM_017014214.1 | Intron | XP_016869703.1 | ||||
XM_017014215.1 | Intron | XP_016869704.1 | ||||
XM_017014216.1 | Intron | XP_016869705.1 |
Set Membership: |
HapMap |