Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604912 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TAF2 - TATA-box binding protein associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003184.3 | Intron | NP_003175.1 | ||||
XM_006716621.3 | Intron | XP_006716684.1 | ||||
XM_011517259.2 | Intron | XP_011515561.1 | ||||
XM_017013779.1 | Intron | XP_016869268.1 | ||||
XM_017013780.1 | Intron | XP_016869269.1 | ||||
XM_017013781.1 | Intron | XP_016869270.1 | ||||
XM_017013782.1 | Intron | XP_016869271.1 | ||||
XM_017013783.1 | Intron | XP_016869272.1 | ||||
XM_017013784.1 | Intron | XP_016869273.1 | ||||
XM_017013785.1 | Intron | XP_016869274.1 | ||||
XM_017013786.1 | Intron | XP_016869275.1 | ||||
XM_017013787.1 | Intron | XP_016869276.1 | ||||
XM_017013788.1 | Intron | XP_016869277.1 | ||||
XM_017013789.1 | Intron | XP_016869278.1 | ||||
XM_017013790.1 | Intron | XP_016869279.1 | ||||
XM_017013791.1 | Intron | XP_016869280.1 |
Set Membership: |
HapMap Validated |