Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGTTCAACTTTTAATGTATATT[A/G]CTGTCTCAACCAAACACCTCAAAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616405 MIM: 106491 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AAK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AAK1 - AP2 associated kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ANXA4 - annexin A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001153.4 | Intron | NP_001144.1 | ||||
NM_001320698.1 | Intron | NP_001307627.1 | ||||
NM_001320700.1 | Intron | NP_001307629.1 | ||||
NM_001320702.1 | Intron | NP_001307631.1 | ||||
XM_017003943.1 | Intron | XP_016859432.1 | ||||
XM_017003944.1 | Intron | XP_016859433.1 |
Set Membership: |
HapMap |