Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAATACTCGGGTTTCTAAGCTCA[C/G]TCTCTCCTGTAATGCAATCCACTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609488 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf47 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C4orf47 - chromosome 4 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCDC110 - coiled-coil domain containing 110 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145411.1 | Intron | NP_001138883.1 | ||||
NM_152775.3 | Intron | NP_689988.1 | ||||
XM_005262891.3 | Intron | XP_005262948.1 | ||||
XM_006714172.2 | Intron | XP_006714235.1 | ||||
XM_011531827.2 | Intron | XP_011530129.1 | ||||
XM_011531828.2 | Intron | XP_011530130.1 | ||||
XM_011531829.2 | Intron | XP_011530131.1 | ||||
XM_017007987.1 | Intron | XP_016863476.1 | ||||
XM_017007988.1 | Intron | XP_016863477.1 |
Set Membership: |
HapMap |