Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM64A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM64A - family with sequence similarity 64 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITPNM3 - PITPNM family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165966.1 | 4916 | UTR 3 | NP_001159438.1 | |||
NM_031220.3 | 4916 | UTR 3 | NP_112497.2 | |||
XM_011524015.2 | 4916 | Intron | XP_011522317.1 | |||
XM_011524016.2 | 4916 | Intron | XP_011522318.1 | |||
XM_011524017.2 | 4916 | Intron | XP_011522319.1 |