Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604566 MIM: 605494 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ALG6 - ALG6, alpha-1,3-glucosyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITGB3BP - integrin subunit beta 3 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206739.1 | Intron | NP_001193668.1 | ||||
NM_014288.4 | Intron | NP_055103.3 | ||||
XM_006710514.3 | Intron | XP_006710577.3 | ||||
XM_006710515.3 | Intron | XP_006710578.3 | ||||
XM_017000853.1 | Intron | XP_016856342.1 | ||||
XM_017000854.1 | Intron | XP_016856343.1 | ||||
XM_017000855.1 | Intron | XP_016856344.1 | ||||
XM_017000856.1 | Intron | XP_016856345.1 | ||||
XM_017000857.1 | Intron | XP_016856346.1 |