Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604939 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLA2R1 - phospholipase A2 receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007267.2 | Intron | NP_001007268.1 | ||||
NM_001195641.1 | Intron | NP_001182570.1 | ||||
NM_007366.4 | Intron | NP_031392.3 | ||||
XM_005246392.3 | Intron | XP_005246449.1 | ||||
XM_011510820.2 | Intron | XP_011509122.1 | ||||
XM_017003598.1 | Intron | XP_016859087.1 | ||||
XM_017003599.1 | Intron | XP_016859088.1 | ||||
XM_017003600.1 | Intron | XP_016859089.1 | ||||
XM_017003601.1 | Intron | XP_016859090.1 | ||||
XM_017003602.1 | Intron | XP_016859091.1 |