Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612218 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB38 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZBTB38 - zinc finger and BTB domain containing 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080412.2 | Intron | NP_001073881.2 | ||||
XM_005247256.4 | Intron | XP_005247313.1 | ||||
XM_005247257.1 | Intron | XP_005247314.1 | ||||
XM_005247258.1 | Intron | XP_005247315.1 | ||||
XM_005247261.3 | Intron | XP_005247318.1 | ||||
XM_011512607.1 | Intron | XP_011510909.1 | ||||
XM_011512608.1 | Intron | XP_011510910.1 | ||||
XM_011512611.1 | Intron | XP_011510913.1 | ||||
XM_017006050.1 | Intron | XP_016861539.1 | ||||
XM_017006051.1 | Intron | XP_016861540.1 | ||||
XM_017006052.1 | Intron | XP_016861541.1 | ||||
XM_017006053.1 | Intron | XP_016861542.1 | ||||
XM_017006054.1 | Intron | XP_016861543.1 | ||||
XM_017006055.1 | Intron | XP_016861544.1 | ||||
XM_017006056.1 | Intron | XP_016861545.1 | ||||
XM_017006057.1 | Intron | XP_016861546.1 | ||||
XM_017006058.1 | Intron | XP_016861547.1 | ||||
XM_017006059.1 | Intron | XP_016861548.1 | ||||
XM_017006060.1 | Intron | XP_016861549.1 | ||||
XM_017006061.1 | Intron | XP_016861550.1 |
Set Membership: |
HapMap |