Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACATGAGAAGTTAGTGAATTAG[A/G]GCCCTCCAAATAAAGATTGTTTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600051 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPS15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EPS15 - epidermal growth factor receptor pathway substrate 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159969.1 | Intron | NP_001153441.1 | ||||
NM_001981.2 | Intron | NP_001972.1 | ||||
XM_005270618.2 | Intron | XP_005270675.1 | ||||
XM_017000615.1 | Intron | XP_016856104.1 | ||||
XM_017000616.1 | Intron | XP_016856105.1 | ||||
XM_017000617.1 | Intron | XP_016856106.1 | ||||
XM_017000618.1 | Intron | XP_016856107.1 | ||||
XM_017000619.1 | Intron | XP_016856108.1 | ||||
XM_017000620.1 | Intron | XP_016856109.1 |
Set Membership: |
HapMap Validated |