Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTCCAGCCAGGAATGGGAAAGG[A/G]GGCTGAAGGTACAGAGGGATAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCOC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCOC - short coiled-coil protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001153446.1 | Intron | NP_001146918.1 | ||||
NM_001153484.1 | Intron | NP_001146956.1 | ||||
NM_001153552.1 | Intron | NP_001147024.1 | ||||
NM_001153585.1 | Intron | NP_001147057.1 | ||||
NM_001153635.1 | Intron | NP_001147107.1 | ||||
NM_001153663.1 | Intron | NP_001147135.1 | ||||
NM_001153690.1 | Intron | NP_001147162.1 | ||||
NM_032547.2 | Intron | NP_115936.2 |