Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATGATCCTGGGGCATCTTGGTG[C/T]AGATCCTGGGAGGCCAGGGCTGCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153432 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LSP1 - lymphocyte-specific protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013253.1 | Intron | NP_001013271.1 | ||||
NM_001013254.1 | Intron | NP_001013272.1 | ||||
NM_001013255.1 | Intron | NP_001013273.1 | ||||
NM_001242932.1 | Intron | NP_001229861.1 | ||||
NM_001289005.1 | Intron | NP_001275934.1 | ||||
NM_002339.2 | Intron | NP_002330.1 |
MIR4298 - microRNA 4298 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |