Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTAGGTCTCAAAATATAACTC[A/G]GCTGTTTCACACTGTATATGTACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604299 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
APPL1 - adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012096.2 | 3858 | UTR 3 | NP_036228.1 | |||
XM_011533583.2 | 3858 | UTR 3 | XP_011531885.1 |
ASB14 - ankyrin repeat and SOCS box containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142733.2 | 3858 | Intron | NP_001136205.2 | |||
NM_130387.5 | 3858 | Intron | NP_569058.1 | |||
XM_017005736.1 | 3858 | Intron | XP_016861225.1 | |||
XM_017005737.1 | 3858 | Intron | XP_016861226.1 |
LOC105377102 - uncharacterized LOC105377102 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |