Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGGGAAGGAGTGGGAGGGCAG[A/G]GGGCAAACTCAGGAACATAGATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602419 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EGR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EGR3 - early growth response 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199880.1 | 2407 | UTR 3 | NP_001186809.1 | |||
NM_001199881.1 | 2407 | UTR 3 | NP_001186810.1 | |||
NM_004430.2 | 2407 | UTR 3 | NP_004421.2 | |||
XM_005273425.3 | 2407 | UTR 3 | XP_005273482.1 | |||
XM_005273426.3 | 2407 | UTR 3 | XP_005273483.1 | |||
XM_011544429.2 | 2407 | UTR 3 | XP_011542731.1 |
Set Membership: |
HapMap |