Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCDC12 - coiled-coil domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277074.1 | 581 | UTR 3 | NP_001264003.1 | |||
NM_144716.5 | 581 | UTR 3 | NP_653317.2 | |||
XM_011533390.2 | 581 | Intron | XP_011531692.1 | |||
XM_011533391.2 | 581 | Intron | XP_011531693.1 | |||
XM_011533392.2 | 581 | Intron | XP_011531694.1 | |||
XM_011533393.2 | 581 | Intron | XP_011531695.1 | |||
XM_011533394.2 | 581 | Intron | XP_011531696.1 | |||
XM_011533396.2 | 581 | UTR 3 | XP_011531698.1 |
Set Membership: |
HapMap Validated |