Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAAAGAAGTATCTGCGCATACAG[C/T]AGCATTATTCACAATAGCCAAATGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610876 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HACE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
HACE1 - HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321080.1 | Intron | NP_001308009.1 | ||||
NM_001321083.1 | Intron | NP_001308012.1 | ||||
NM_001321084.1 | Intron | NP_001308013.1 | ||||
NM_020771.3 | Intron | NP_065822.2 | ||||
XM_017011113.1 | Intron | XP_016866602.1 | ||||
XM_017011114.1 | Intron | XP_016866603.1 | ||||
XM_017011115.1 | Intron | XP_016866604.1 | ||||
XM_017011116.1 | Intron | XP_016866605.1 | ||||
XM_017011118.1 | Intron | XP_016866607.1 | ||||
XM_017011119.1 | Intron | XP_016866608.1 | ||||
XM_017011120.1 | Intron | XP_016866609.1 | ||||
XM_017011121.1 | Intron | XP_016866610.1 | ||||
XM_017011122.1 | Intron | XP_016866611.1 |
Set Membership: |
HapMap |