Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176949 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAPK4 - mitogen-activated protein kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292039.1 | Intron | NP_001278968.1 | ||||
NM_001292040.1 | Intron | NP_001278969.1 | ||||
NM_002747.3 | Intron | NP_002738.2 | ||||
XM_005258299.3 | Intron | XP_005258356.1 | ||||
XM_011526074.2 | Intron | XP_011524376.1 | ||||
XM_011526075.2 | Intron | XP_011524377.1 | ||||
XM_011526076.2 | Intron | XP_011524378.1 | ||||
XM_011526077.2 | Intron | XP_011524379.1 | ||||
XM_011526078.2 | Intron | XP_011524380.1 | ||||
XM_017025839.1 | Intron | XP_016881328.1 |
Set Membership: |
HapMap |