Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGGCCACCCGGTGAAGAGGGAA[A/C]CGATTTCTCTGTGGGGCTTCAGAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC11C PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEC11C - SEC11 homolog C, signal peptidase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307941.1 | Intron | NP_001294870.1 | ||||
NM_033280.3 | Intron | NP_150596.1 | ||||
XM_011526260.2 | Intron | XP_011524562.1 | ||||
XM_017026073.1 | Intron | XP_016881562.1 |
Set Membership: |
HapMap |