Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTGGACGTGAGACATATCCCAG[A/G]AGGTTGAATTTGGAAATACCAGTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137260 | ||||||||||||||||||||
Literature Links: |
GRP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRP - gastrin releasing peptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012512.2 | Intron | NP_001012530.1 | ||||
NM_001012513.2 | Intron | NP_001012531.1 | ||||
NM_002091.4 | Intron | NP_002082.2 | ||||
XM_011525933.2 | Intron | XP_011524235.1 | ||||
XM_017025712.1 | Intron | XP_016881201.1 | ||||
XM_017025713.1 | Intron | XP_016881202.1 | ||||
XM_017025714.1 | Intron | XP_016881203.1 |