Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAACTCATTCTTACGACACCCTC[A/G]TTGCCCTAATGCCAAAACCAGACCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605491 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf113 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C10orf113 - chromosome 10 open reading frame 113 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010896.2 | Intron | NP_001010896.2 | ||||
NM_001177483.1 | Intron | NP_001170954.1 |
NEBL - nebulette | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173484.1 | Intron | NP_001166955.1 | ||||
NM_006393.2 | Intron | NP_006384.1 | ||||
NM_213569.2 | Intron | NP_998734.1 | ||||
XM_005252342.4 | Intron | XP_005252399.1 | ||||
XM_005252343.4 | Intron | XP_005252400.1 | ||||
XM_005252344.4 | Intron | XP_005252401.1 | ||||
XM_011519291.2 | Intron | XP_011517593.1 | ||||
XM_017015467.1 | Intron | XP_016870956.1 | ||||
XM_017015468.1 | Intron | XP_016870957.1 | ||||
XM_017015469.1 | Intron | XP_016870958.1 |