Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614517 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARNTL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ARNTL2 - aryl hydrocarbon receptor nuclear translocator like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001248002.1 | Intron | NP_001234931.1 | ||||
NM_001248003.1 | Intron | NP_001234932.1 | ||||
NM_001248004.1 | Intron | NP_001234933.1 | ||||
NM_001248005.1 | Intron | NP_001234934.1 | ||||
NM_020183.4 | Intron | NP_064568.3 | ||||
XM_006719112.3 | Intron | XP_006719175.1 | ||||
XM_006719114.3 | Intron | XP_006719177.1 | ||||
XM_011520766.2 | Intron | XP_011519068.1 | ||||
XM_011520768.2 | Intron | XP_011519070.1 | ||||
XM_011520769.2 | Intron | XP_011519071.1 | ||||
XM_011520770.2 | Intron | XP_011519072.1 | ||||
XM_011520771.2 | Intron | XP_011519073.1 | ||||
XM_017019666.1 | Intron | XP_016875155.1 | ||||
XM_017019667.1 | Intron | XP_016875156.1 | ||||
XM_017019668.1 | Intron | XP_016875157.1 | ||||
XM_017019669.1 | Intron | XP_016875158.1 |
Set Membership: |
HapMap Validated |