Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAGACACTCAAGCAGAGGCCAG[C/G]TGGCCTTGAAGTATGATTCCATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611156 MIM: 610354 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERMP1 - endoplasmic reticulum metallopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024896.2 | Intron | NP_079172.2 | ||||
XM_005251587.1 | Intron | XP_005251644.1 | ||||
XM_017015139.1 | Intron | XP_016870628.1 |
RIC1 - RIC1 homolog, RAB6A GEF complex partner 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135920.2 | Intron | NP_001129392.2 | ||||
NM_001206557.1 | Intron | NP_001193486.1 | ||||
NM_020829.3 | Intron | NP_065880.2 | ||||
XM_005251523.3 | Intron | XP_005251580.1 | ||||
XM_011517967.2 | Intron | XP_011516269.1 | ||||
XM_011517968.2 | Intron | XP_011516270.1 | ||||
XM_017014934.1 | Intron | XP_016870423.1 | ||||
XM_017014935.1 | Intron | XP_016870424.1 | ||||
XM_017014936.1 | Intron | XP_016870425.1 |