Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608925 MIM: 602998 | ||||||||||||||||||||||||||||||||
Literature Links: |
ADIRF PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ADIRF - adipogenesis regulatory factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AGAP11 - ArfGAP with GTPase domain, ankyrin repeat and PH domain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133447.1 | Intron | NP_597704.1 |
MMRN2 - multimerin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024756.2 | Intron | NP_079032.2 | ||||
XM_005270153.1 | Intron | XP_005270210.1 | ||||
XM_006717970.3 | Intron | XP_006718033.1 |
SNCG - synuclein gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |