Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCAGGTTCCCCAGACTTGTTGAT[A/G]TAATTAGAAATTGAATGTCAGCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603888 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNS3 - potassium voltage-gated channel modifier subfamily S member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282428.1 | Intron | NP_001269357.1 | ||||
NM_002252.4 | Intron | NP_002243.3 | ||||
XM_011532825.1 | Intron | XP_011531127.1 | ||||
XM_017004059.1 | Intron | XP_016859548.1 |