Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGAGACCTTTCCCAACCCCTGT[T/C]TCCACCTCTGTGTCTGCAGCTGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
73 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605548 MIM: 601380 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADAM15 - ADAM metallopeptidase domain 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFNA4 - ephrin A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005227.2 | Intron | NP_005218.1 | ||||
NM_182689.1 | Intron | NP_872631.1 | ||||
NM_182690.2 | Intron | NP_872632.2 |
LOC100505666 - uncharacterized LOC100505666 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |