Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC37A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SLC37A2 - solute carrier family 37 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM218 - transmembrane protein 218 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080546.2 | 995 | UTR 3 | NP_001074015.1 | |||
NM_001258238.1 | 995 | UTR 3 | NP_001245167.1 | |||
NM_001258239.1 | 995 | UTR 3 | NP_001245168.1 | |||
NM_001258240.1 | 995 | UTR 3 | NP_001245169.1 | |||
NM_001258241.1 | 995 | UTR 3 | NP_001245170.1 | |||
NM_001258242.1 | 995 | UTR 3 | NP_001245171.1 | |||
NM_001258243.1 | 995 | UTR 3 | NP_001245172.1 | |||
NM_001258244.1 | 995 | UTR 3 | NP_001245173.1 | |||
NM_001258245.1 | 995 | UTR 3 | NP_001245174.1 | |||
NM_001258246.1 | 995 | UTR 3 | NP_001245175.1 | |||
NM_001258247.1 | 995 | UTR 3 | NP_001245176.1 | |||
XM_006718779.3 | 995 | UTR 3 | XP_006718842.1 | |||
XM_006718780.3 | 995 | UTR 3 | XP_006718843.1 | |||
XM_006718781.3 | 995 | UTR 3 | XP_006718844.1 | |||
XM_006718783.3 | 995 | UTR 3 | XP_006718846.1 | |||
XM_017017322.1 | 995 | UTR 3 | XP_016872811.1 | |||
XM_017017323.1 | 995 | UTR 3 | XP_016872812.1 | |||
XM_017017324.1 | 995 | UTR 3 | XP_016872813.1 |