Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAAAAGCAATTATATGTCATTAG[A/G]TAGATGAGGTTACTTTTGTGAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609211 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC104968399 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC104968399 - uncharacterized LOC104968399 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYL12A - myosin light chain 12A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYL12B - myosin light chain 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144944.1 | Intron | NP_001138416.1 | ||||
NM_001144945.1 | Intron | NP_001138417.1 | ||||
NM_033546.3 | Intron | NP_291024.1 |
Set Membership: |
HapMap Validated |