Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611909 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC3B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FNDC3B - fibronectin type III domain containing 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135095.1 | Intron | NP_001128567.1 | ||||
NM_022763.3 | Intron | NP_073600.3 | ||||
XM_017007062.1 | Intron | XP_016862551.1 | ||||
XM_017007063.1 | Intron | XP_016862552.1 | ||||
XM_017007064.1 | Intron | XP_016862553.1 |
Set Membership: |
HapMap |