Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614451 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELOVL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ELOVL7 - ELOVL fatty acid elongase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104558.1 | Intron | NP_001098028.1 | ||||
NM_001297617.1 | Intron | NP_001284546.1 | ||||
NM_001297618.1 | Intron | NP_001284547.1 | ||||
NM_024930.2 | Intron | NP_079206.2 | ||||
XM_005248606.4 | Intron | XP_005248663.1 | ||||
XM_005248607.3 | Intron | XP_005248664.1 | ||||
XM_006714695.3 | Intron | XP_006714758.1 | ||||
XM_011543651.2 | Intron | XP_011541953.1 | ||||
XM_017009885.1 | Intron | XP_016865374.1 | ||||
XM_017009886.1 | Intron | XP_016865375.1 | ||||
XM_017009887.1 | Intron | XP_016865376.1 | ||||
XM_017009888.1 | Intron | XP_016865377.1 | ||||
XM_017009889.1 | Intron | XP_016865378.1 |
Set Membership: |
HapMap Validated |