Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGAGAGAGTGAAACGGGGGAGC[A/C]GGCTACATCACCCACAGCCTGATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604182 MIM: 608855 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGC16275 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MGC16275 - uncharacterized protein MGC16275 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL38 - ribosomal protein L38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTYH2 - tweety family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032646.5 | Intron | NP_116035.5 | ||||
NM_052869.1 | Intron | NP_443101.1 | ||||
XM_005257824.3 | Intron | XP_005257881.1 |
Set Membership: |
HapMap |