Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609585 MIM: 606912 MIM: 613472 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPLX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CPLX3 - complexin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6882 - microRNA 6882 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCAMP2 - secretory carrier membrane protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ULK3 - unc-51 like kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099436.3 | 2646 | UTR 3 | NP_001092906.3 | |||
NM_001284364.2 | 2646 | UTR 3 | NP_001271293.2 | |||
NM_001284365.2 | 2646 | UTR 3 | NP_001271294.1 | |||
XM_005254289.2 | 2646 | UTR 3 | XP_005254346.1 | |||
XM_017022068.1 | 2646 | UTR 3 | XP_016877557.1 | |||
XM_017022069.1 | 2646 | UTR 3 | XP_016877558.1 | |||
XM_017022070.1 | 2646 | UTR 3 | XP_016877559.1 | |||
XM_017022071.1 | 2646 | UTR 3 | XP_016877560.1 | |||
XM_017022072.1 | 2646 | UTR 3 | XP_016877561.1 |
Set Membership: |
HapMap |